4,8-双(羟甲基)三环[5.2.1.02,6]癸烷,异构体混合物 ≥90.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
三环[5.2.1.02,6]癸烷-4,8-二甲醇
T0850-25G
T0850-500G
TCIAT0850-25GEA
0
CNY
TCIAT0850-25G
TCIAT0850-500G
4,8-双(羟甲基)三环[5.2.1.02,6]癸烷,异构体混合物 ≥90.0% (by GC)
4,8-双(羟甲基)三环[5.2.1.02,6]癸烷,异构体混合物
分子式:
C₁₂H₂₀O₂ 分子量: 196.29 g/mol 闪点: 185°C |
MDL编号:
MFCD00151166 CAS号: 26896-48-0 EINECS: 248-096-5 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0% (total of diastereoisomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...