Thiopropionamide ≥98.0% (通过GC)

供应商: TCI
Warning

T0669-10G T0669-25G
TCIAT0669-10GEA 0 CNY
TCIAT0669-10G TCIAT0669-25G
Thiopropionamide ≥98.0% (通过GC)
Thiopropionamide
分子式: C₃H₇NS
存储条件: Refrigerator
MDL编号: MFCD00059864
CAS号: 631-58-3

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 42.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR