三丙二醇 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Tripropylene glycol

T0523-25ML T0523-500ML
TCIAT0523-25MLEA 0 CNY
TCIAT0523-25ML TCIAT0523-500ML
三丙二醇 ≥98.0% (by GC)
三丙二醇
分子式: C₉H₂₀O₄
分子量: 192.26 g/mol
闪点: 146°C
MDL编号: MFCD00014405
CAS号: 24800-44-0
EINECS: 246-466-0

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR