Tridecanol (mixture of isomers)
供应商: TCI
|
Warning
|
Synonyms:
Tridecyl Alcohol
T0414-25ML
T0414-500ML
TCIAT0414-25MLEA
0
CNY
TCIAT0414-25ML
TCIAT0414-500ML
Tridecanol (mixture of isomers)
Tridecanol (mixture of isomers)
分子式:
C₁₃H₂₈O 分子量: 200.37 g/mol 闪点: 130°C |
MDL编号:
MFCD01778419 CAS号: 26248-42-0 |
产品技术规格
Appearance | Colorless clear liquid |
Purity (GC) | many component mixture |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...