o-Toluidine-4-sulphonic acid sodium salt tetrahydrate ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 2-Aminotoluene-5-sulphonic acid Sodium Salt, Sodium 4-Amino-3-methylbenzenesulphonate, 4-Amino-3-methylbenzenesulphonic acid Sodium Salt, Sodium 2-Aminotoluene-5-sulphonate, Sodium o-Toluidine-4-sulphonate

T0304-25G T0304-500G
TCIAT0304-25GEA 0 CNY
TCIAT0304-25G TCIAT0304-500G
o-Toluidine-4-sulphonic acid sodium salt tetrahydrate ≥98.0% (by titrimetric analysis)
o-Toluidine-4-sulphonic acid sodium salt tetrahydrate
分子式: C₇H₉NO₃S
分子量: 209.19 g/mol
MDL编号: MFCD00070549
CAS号: 63450-43-1
EINECS: 264-172-0

现在订购

产品技术规格

Purity (Ion exchange titration) min. 98.0 % (calcd.on anh.substance)
Water 25.0 to 26.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR