Tetraiodo-2-sulphobenzoic anhydride ≥95.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
T0131-1G
T0131-25G
TCIAT0131-1GEA
0
CNY
TCIAT0131-1G
TCIAT0131-25G
Tetraiodo-2-sulphobenzoic anhydride ≥95.0% (by titrimetric analysis)
Tetraiodo-2-sulphobenzoic anhydride
分子式:
C₇I₄O₄S 分子量: 687.75 g/mol |
MDL编号:
MFCD00059713 CAS号: 1745-83-1 UN号: 1759 ADR: 8 III |
产品技术规格
Appearance | Light yellow to Yellow powder to crystal |
Purity (Methanolysis Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...