7,7,8,8-四氰基对苯二醌二甲烷 ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
|
Danger
|
Synonyms:
TCNQ, 2,2'-(2,5-环己二烯-1,4-二基)丙二腈
T0078-25G
T0078-5G
TCIAT0078-25GEA
0
CNY
TCIAT0078-25G
TCIAT0078-5G
7,7,8,8-四氰基对苯二醌二甲烷 ≥98.0% (by HPLC, total nitrogen)
7,7,8,8-四氰基对苯二醌二甲烷
分子式:
C₁₂H₄N₄ |
MDL编号:
MFCD00011664 CAS号: 1518-16-7 EINECS: 216-174-8 UN号: 3439 ADR: 6.1,III |
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Acetonitrile | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...