DL-酒石酸 ≥99.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
DL-2,3-二羟基丁二酸
T0001-25G
T0001-500G
TCIAT0001-25GEA
0
CNY
TCIAT0001-25G
TCIAT0001-500G
DL-酒石酸 ≥99.0% (by GC, titration analysis)
DL-酒石酸
分子式:
C₄H₆O₆ 分子量: 150.09 g/mol |
MDL编号:
MFCD00151254 CAS号: 133-37-9 EINECS: 205-105-7 Merck Index: 14,09069 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...