Sialyl lewis-X-lactose ≥97.0% (by HPLC)

供应商: TCI

Synonyms: Neu5Ac alpha(2-3)Gal beta(1-4)[Fuc alpha(1-3)]GlcNAc beta(1-3)Gal beta(1-4)Glc

S0849-1MG S0849-5MG
TCIAS0849-1MGEA 0 CNY
TCIAS0849-1MG TCIAS0849-5MG
Sialyl lewis-X-lactose ≥97.0% (by HPLC)
Sialyl lewis-X-lactose
分子式: C₄₃H₇₂N₂O₃₃
分子量: 1145.03 g/mol
存储条件: Freezer
CAS号: 127923-85-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR