Sodium-3-(N-ethylanilino)propanesulphonate ≥98.0% (by HPLC) for biochemical research

供应商: TCI

Synonyms: N-Ethyl-N-(3-sulphopropyl)aniline Sodium Salt, ALPS, 3-(N-Ethylanilino)propanesulphonic acid Sodium Salt, Sodium 3-(ethyl(phenyl)amino)propane-1-sulphonate

S0817-1G S0817-200MG
TCIAS0817-1GEA 0 CNY
TCIAS0817-1G TCIAS0817-200MG
Sodium-3-(N-ethylanilino)propanesulphonate ≥98.0% (by HPLC) for biochemical research
Sodium-3-(N-ethylanilino)propanesulphonate
分子式: C₁₁H₁₇NO₃S
分子量: 265.30 g/mol
MDL编号: MFCD01311047
CAS号: 82611-85-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Hydrogendioxide detection test (Peroxidase method) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR