1,1′-磺酰二咪唑 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
S0803-25G
S0803-5G
TCIAS0803-25GEA
0
CNY
TCIAS0803-25G
TCIAS0803-5G
1,1′-磺酰二咪唑 ≥98.0% (通过GC)
1,1′-磺酰二咪唑
分子式:
C₆H₆N₄O₂S |
MDL编号:
MFCD00015893 CAS号: 7189-69-7 EINECS: 230-554-0 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...