Sodium cholate (from bovine bile) ≥98.0% (by titrimetric analysis) for biochemical research

供应商: TCI

Synonyms: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt

S0596-5G S0596-25G
TCIAS0596-5GEA 0 CNY
TCIAS0596-5G TCIAS0596-25G
Sodium cholate (from bovine bile) ≥98.0% (by titrimetric analysis) for biochemical research
Sodium cholate
分子式: C₂₄H₃₉NaO₅
分子量: 430.56 g/mol
溶点: 198
MDL编号: MFCD00150749
CAS号: 361-09-1
EINECS: 206-643-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (after drying)
Specific rotation [a]20/D +34.0 to +36.0 deg (C=0.6, Ethanol)
Solubility in Water almost transparency
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR