磺胺二甲基嘧啶 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 4,6-二甲基磺胺嘧啶, 4-氨基-N-(4,6-二甲基-2-嘧啶基)苯磺酰胺, 磺胺二甲嘧啶

S0586-25G S0586-250G
TCIAS0586-25GEA 0 CNY
TCIAS0586-25G TCIAS0586-250G
磺胺二甲基嘧啶 ≥98.0% (by HPLC, titration analysis)
磺胺二甲基嘧啶
分子式: C₁₂H₁₄N₄O₂S
存储条件: Refrigerator
MDL编号: MFCD00006066
CAS号: 57-68-1
EINECS: 200-346-4
Merck Index: 14,08905

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR