Sulphacetamide sodium salt ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: N-Acetyl-4-aminobenzenesulphonamide sodium salt, N-Acetylsulphanilamide sodium salt

S0578-25G S0578-250G
TCIAS0578-25GEA 0 CNY
TCIAS0578-25G TCIAS0578-250G
Sulphacetamide sodium salt ≥98.0% (by HPLC, titration analysis)
Sulphacetamide sodium salt
分子式: C₈H₁₀N₂O₃S
分子量: 236.22 g/mol
MDL编号: MFCD00007885
CAS号: 127-56-0
EINECS: 204-848-4
Merck Index: 14,08899

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR