Santonin ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: (3S,3aS,5aS,9bS)-3a,5,5a,9b-Tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)-dione

S0521-25G S0521-5G
TCIAS0521-25GEA 0 CNY
TCIAS0521-25G TCIAS0521-5G
Santonin ≥98.0% (by HPLC, titration analysis)
Santonin
分子式: C₁₅H₁₈O₃
CAS号: 481-06-1
EINECS: 207-560-7
Merck Index: 14,08361

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ester Value) min. 98.0 %
Melting point 172.0 to 175.0 deg-C
Specific rotation [a]20/D -170 to -175 deg (C=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR