Sodium-4-(2-bromoethyl)benzenesulphonate ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 4-(2-Bromoethyl)benzenesulphonic acid Sodium Salt

S0484-25G S0484-5G
TCIAS0484-25GEA 0 CNY
TCIAS0484-25G TCIAS0484-5G
Sodium-4-(2-bromoethyl)benzenesulphonate ≥98.0% (by titrimetric analysis)
Sodium-4-(2-bromoethyl)benzenesulphonate
分子式: C₈H₉BrO₃S
分子量: 287.10 g/mol
MDL编号: MFCD06797167
CAS号: 65036-65-9
EINECS: 265-318-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR