双(2,2,6,6-四甲基-4-哌啶基)癸二酸酯 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Sebacic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester, Bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate, Decanedioic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester
S0448-25G
S0448-250G
S0448-5G
TCIAS0448-25GEA
0
CNY
TCIAS0448-25G
TCIAS0448-250G
TCIAS0448-5G
双(2,2,6,6-四甲基-4-哌啶基)癸二酸酯 ≥98.0% (by GC)
双(2,2,6,6-四甲基-4-哌啶基)癸二酸酯
分子式:
C₂₈H₅₂N₂O₄ 分子量: 480.73 g/mol |
MDL编号:
MFCD00134709 CAS号: 52829-07-9 EINECS: 258-207-9 UN号: 2811 ADR: 6.1 II |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...