Iron(III) stearate

供应商: TCI

Synonyms: Ferric Stearate, Iron(III) Stearate, Stearic Acid Iron(III) Salt

S0256-25G S0256-500G
TCIAS0256-25GEA 0 CNY
TCIAS0256-25G TCIAS0256-500G
Iron(III) stearate
Iron(III) stearate
分子式: C₁₈H₃₆O₂
分子量: 906.27 g/mol
MDL编号: MFCD00050716
CAS号: 555-36-2
EINECS: 209-095-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Stearic acid (composition of fatty acid) min. 60.0 %
Iron 5.8 to 7.0 %
Free Acid max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR