1,3-苯二甲酸二甲酯-5-磺酸 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI
Warning

S0157-25G S0157-500G
TCIAS0157-25GEA 0 CNY
TCIAS0157-25G TCIAS0157-500G
1,3-苯二甲酸二甲酯-5-磺酸 ≥98.0% (通过HPLC,滴定分析)
1,3-苯二甲酸二甲酯-5-磺酸
分子式: C₁₀H₁₀O₇S
MDL编号: MFCD00007493
CAS号: 3965-55-7
EINECS: 223-578-8

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR