Sodium sorbate ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Sorbic Acid Sodium Salt

S0058-25G S0058-500G
TCIAS0058-25GEA 0 CNY
TCIAS0058-25G TCIAS0058-500G
Sodium sorbate ≥98.0% (by titrimetric analysis)
Sodium sorbate
分子式: C₆H₈O₂
MDL编号: MFCD00058995
CAS号: 7757-81-5
EINECS: 231-819-3

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR