水杨酸异丁酯 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Hydroxybenzoic Acid Isobutyl Ester, Salicylic Acid Isobutyl Ester
S0013-25G
S0013-500G
TCIAS0013-25GEA
0
CNY
TCIAS0013-25G
TCIAS0013-500G
水杨酸异丁酯 ≥99.0% (by GC)
水杨酸异丁酯
分子式:
C₁₁H₁₄O₃ 分子量: 194.23 g/mol 沸点: 259°C 闪点: 121°C |
MDL编号:
MFCD00020035 CAS号: 87-19-4 EINECS: 201-729-9 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...