水杨酸异戊酯 (contains 2-Methylbutyl Salicylate)
供应商: TCI
|
Warning
|
Synonyms:
Isopentyl 2-Hydroxybenzoate, Isopentyl Salicylate, Salicylic Acid Isoamyl Ester, Isopentyl 2-hydroxybenzoate
S0012-25ML
S0012-500ML
TCIAS0012-25MLEA
0
CNY
TCIAS0012-25ML
TCIAS0012-500ML
水杨酸异戊酯 (contains 2-Methylbutyl Salicylate)
水杨酸异戊酯
分子式:
C₁₂H₁₆O₃ 分子量: 208.26 g/mol 沸点: 273°C 闪点: 146°C |
MDL编号:
MFCD00020037 CAS号: 87-20-7 EINECS: 201-730-4 UN号: 3082 ADR: 9 III Merck Index: 14,05125 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...