D-(+)-核糖酸 γ-内酯 ≥97.0% (by GC)

供应商: TCI

Synonyms: D-(+)-核糖酸-1,4-内酯

R0063-1G R0063-5G
TCIAR0063-1GEA 0 CNY
TCIAR0063-1G TCIAR0063-5G
D-(+)-核糖酸 γ-内酯 ≥97.0% (by GC)
D-(+)-核糖酸 γ-内酯
分子式: C₅H₈O₅
分子量: 148.11 g/mol
MDL编号: MFCD00063241
CAS号: 5336-08-3
EINECS: 226-256-5

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Specific rotation [a]20/D +15.0 to +20.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR