喹啉-N-氧化物 ≥97.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Quinoline 1-oxide hydrate

Q0072-25G Q0072-5G
TCIAQ0072-25GEA 0 CNY
TCIAQ0072-25G TCIAQ0072-5G
喹啉-N-氧化物 ≥97.0% (by titrimetric analysis)
喹啉-N-氧化物
分子式: C₉H₇NO·xH₂O
分子量: 145.16 g/mol (anhy)
沸点: 173°C
MDL编号: MFCD00149472
CAS号: 198878-42-1
EINECS: 216-560-6

现在订购

产品技术规格

Appearance White to Brown powder to crystal
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Water 15.0 to 21.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR