Quinaldic acid sodium salt ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
Sodium 2-Quinolinecarboxylate, Sodium Quinaldinate, 2-Quinolinecarboxylic Acid Sodium Salt
Q0004-1G
Q0004-25G
TCIAQ0004-1GEA
0
CNY
TCIAQ0004-1G
TCIAQ0004-25G
Quinaldic acid sodium salt ≥98.0% (by HPLC, titration analysis)
Quinaldic acid sodium salt
分子式:
C₁₀H₇NO₂ 分子量: 195.15 g/mol |
MDL编号:
MFCD00070526 CAS号: 16907-79-2 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...