Paroxetine hydrochloride hemihydrate ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
(3S,4R)-(-)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine Hydrochloride
P1977-1G
P1977-5G
TCIAP1977-1GEA
0
CNY
TCIAP1977-1G
TCIAP1977-5G
Paroxetine hydrochloride hemihydrate ≥98.0% (by HPLC, titration analysis)
Paroxetine hydrochloride hemihydrate
分子式:
C₁₉H₂₀FNO₃ 分子量: 365.83 g/mol |
MDL编号:
MFCD03658863 CAS号: 110429-35-1 Merck Index: 14,07043 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -88.0 to -92.0 deg (C=1, MeOH, calcd.on anh.substance) |
Water | 2.0 to 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...