(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: (4S,4'S)-2,2'-(pentane-3,3-diyl)bis(4-benzyl-4,5-dihydrooxazole), (S,S)-3,3-Bis(4-benzyl-2-oxazolin-2-yl)pentane, (S,S)-2,2'-(Diethylmethylene)bis(4-benzyl-2-oxazoline)

P1947-1G P1947-200MG
TCIAP1947-1GEA 0 CNY
TCIAP1947-1G TCIAP1947-200MG
(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥98.0% (by titrimetric analysis)
(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole)
分子式: C₂₅H₃₀N₂O₂
分子量: 390.53 g/mol
MDL编号: MFCD20265601
CAS号: 160191-64-0

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -20.0 to -23.0 deg (C=2.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR