三氟乙酸五氟苯酯 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: 五氟苯基三氟乙酸酯

P1894-25G P1894-5G
TCIAP1894-25GEA 0 CNY
TCIAP1894-25G TCIAP1894-5G
三氟乙酸五氟苯酯 ≥95.0% (by GC)
三氟乙酸五氟苯酯
分子式: C₈F₈O₂
分子量: 280.07 g/mol
沸点: 122°C
溶点: 4°C
闪点: 52°C
MDL编号: MFCD00134438
CAS号: 14533-84-7
UN号: 3272
ADR: 3 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR