2-Phosphonobutane-1,2,4-tricarboxylic acid ∼50% in water

供应商: TCI
Danger

Synonyms: 3-Carboxy-3-phosphonohexanedioic acid, PBTC

P1857-25G P1857-500G
TCIAP1857-25GEA 0 CNY
TCIAP1857-25G TCIAP1857-500G
2-Phosphonobutane-1,2,4-tricarboxylic acid ∼50% in water
2-Phosphonobutane-1,2,4-tricarboxylic acid
分子式: C₇H₁₁O₉P
MDL编号: MFCD01940753
CAS号: 37971-36-1
EINECS: 253-733-5
UN号: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7822c405b0f92e490b3541c2619d07d178fd6c1&vid=a15f50e07f87fed2eaf3ea4b5ab977aba199af90&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Concentration (Neutralization titration) 49.0 to 51.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR