Phytosphingosine ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: (2S,3S,4R)-2-Aminooctadecane-1,3,4-triol, (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol

P1765-1G P1765-5G
TCIAP1765-1GEA 0 CNY
TCIAP1765-1G TCIAP1765-5G
Phytosphingosine ≥98.0% (by titrimetric analysis)
Phytosphingosine
分子式: C₁₈H₃₉NO₃
溶点: 102°C
MDL编号: MFCD00079232
CAS号: 554-62-1

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +6.0 to +12.0 deg (C=1, Pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR