[N-[ɑ-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel ≥93.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
P1737-1G
P1737-100MG
TCIAP1737-1GEA
0
CNY
TCIAP1737-1G
TCIAP1737-100MG
[N-[ɑ-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel ≥93.0% (by titrimetric analysis)
[N-[alpha-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel
分子式:
C₂₂H₂₃N₃NiO₃ 分子量: 436.14 g/mol |
CAS号:
847654-17-5 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...