Pentadecyl acetate ≥95.0% (by GC)

供应商: TCI

Synonyms: Acetic Acid Pentadecyl Ester

P1700-25G P1700-5G
TCIAP1700-25GEA 0 CNY
TCIAP1700-25G TCIAP1700-5G
Pentadecyl acetate ≥95.0% (by GC)
Pentadecyl acetate
分子式: C₁₇H₃₄O₂
沸点: 230°C
溶点: 15°C
MDL编号: MFCD00056204
CAS号: 629-58-3

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR