1-苯基-1-硫代-β-D-吡喃葡萄糖苷四乙酸 ≥98.0% (by HPLC)

供应商: TCI

Synonyms: 1-Phenyl-1-thio-β-D-glucopyranoside tetraacetate, (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate, Phenyl 2,3,4,6-tetra-o-acetyl-1-thio-β-D-glucopyranoside

P1476-25G P1476-5G
TCIAP1476-25GEA 0 CNY
TCIAP1476-25G TCIAP1476-5G
1-苯基-1-硫代-β-D-吡喃葡萄糖苷四乙酸 ≥98.0% (by HPLC)
1-苯基-1-硫代-β-D-吡喃葡萄糖苷四乙酸
分子式: C₂₀H₂₄O₉S
分子量: 440.46 g/mol
溶点: 118°C
存储条件: Freezer
MDL编号: MFCD00135159
CAS号: 23661-28-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D -15.0 to -17.0 deg (C=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR