2-苯基-1,3-二氧六环-5-醇 ≥90.0% (by NMR)

供应商: TCI

Synonyms: 1,3-O-亚苄基甘油

P1422-1G P1422-5G
TCIAP1422-1GEA 0 CNY
TCIAP1422-1G TCIAP1422-5G
2-苯基-1,3-二氧六环-5-醇 ≥90.0% (by NMR)
2-苯基-1,3-二氧六环-5-醇
分子式: C₁₀H₁₂O₃
存储条件: Refrigerator
MDL编号: MFCD00047504
CAS号: 1708-40-3
EINECS: 216-963-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (NMR) min. 90.0 atom%
Melting point 80.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR