1-Piperidinepentanol ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
1-(5-Hydroxypentyl)piperidine
P1385-1G
P1385-5G
TCIAP1385-1GEA
0
CNY
TCIAP1385-1G
TCIAP1385-5G
1-Piperidinepentanol ≥98.0% (by GC, titration analysis)
1-Piperidinepentanol
分子式:
C₁₀H₂₁NO 分子量: 171.28 g/mol 沸点: 140°C |
MDL编号:
MFCD03093640 CAS号: 2937-83-9 |
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...