(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol ≥97.0% (通过GC)

供应商: TCI
Warning

P1375-1G P1375-5G
TCIAP1375-1GEA 0 CNY
TCIAP1375-1G TCIAP1375-5G
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol ≥97.0% (通过GC)
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol
分子式: C₁₃H₁₉NO
MDL编号: MFCD02093492
CAS号: 123620-80-4

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 42.0 to 48.0 deg-C
Specific rotation [a]20/D -13.0 to -16.0 deg (C=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR