焦碳酸二叔戊酯 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
二碳酸二叔戊酯, (Amoc)2O, 二叔戊基焦碳酸酯
P1356-25G
P1356-5G
TCIAP1356-25GEA
0
CNY
TCIAP1356-25G
TCIAP1356-5G
焦碳酸二叔戊酯 ≥95.0% (by GC)
焦碳酸二叔戊酯
分子式:
C₁₂H₂₂O₅ 沸点: 71°C 闪点: 104°C 存储条件: Refrigerator |
MDL编号:
MFCD00038320 CAS号: 68835-89-2 EINECS: 272-436-1 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...