Methyl-5-phenylvalerate ≥98.0% (by GC)

供应商: TCI

Synonyms: 5-Phenylvaleric Acid Methyl Ester, Methyl 5-phenylpentanoate

P1324-25G P1324-5G
TCIAP1324-25GEA 0 CNY
TCIAP1324-25G TCIAP1324-5G
Methyl-5-phenylvalerate ≥98.0% (by GC)
Methyl-5-phenylvalerate
分子式: C₁₂H₁₆O₂
分子量: 192.26 g/mol
沸点: 173°C
MDL编号: MFCD00025896
CAS号: 20620-59-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR