9-Phenylxanthen-9-ol

供应商: TCI

P1269-25G P1269-5G
TCIAP1269-25GEA 0 CNY
TCIAP1269-25G TCIAP1269-5G
9-Phenylxanthen-9-ol
9-Phenylxanthen-9-ol
分子式: C₁₉H₁₄O₂
MDL编号: MFCD00005058
CAS号: 596-38-3
EINECS: 209-882-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR