Phenyl trimethylsilylmethyl sulphone ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: (Phenylsulphonylmethyl)trimethylsilane

P1250-1G P1250-5G
TCIAP1250-1GEA 0 CNY
TCIAP1250-1G TCIAP1250-5G
Phenyl trimethylsilylmethyl sulphone ≥95.0% (by GC)
Phenyl trimethylsilylmethyl sulphone
分子式: C₁₀H₁₆O₂SSi
MDL编号: MFCD00012364
CAS号: 17872-92-3

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR