7-Ethoxy-7-oxoheptanoic acid ≥98.0% (通过GC)

供应商: TCI
Warning

P1244-25G P1244-5G
TCIAP1244-25GEA 0 CNY
TCIAP1244-25G TCIAP1244-5G
7-Ethoxy-7-oxoheptanoic acid ≥98.0% (通过GC)
分子式: C₉H₁₆O₄
沸点: 162°C
存储条件: Refrigerator
MDL编号: MFCD00801202
CAS号: 33018-91-6
EINECS: 251-346-6

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR