N-(2-Pyridylmethyl)glycine ethyl ester ≥95.0% (by GC)

供应商: TCI

Synonyms: N-(2-Picolyl)glycine Ethyl Ester, Ethyl N-(2-Pyridylmethyl)glycinate, Ethyl 2-((pyridin-2-ylmethyl)amino)acetate

P1194-1G P1194-5G
TCIAP1194-1GEA 0 CNY
TCIAP1194-1G TCIAP1194-5G
N-(2-Pyridylmethyl)glycine ethyl ester ≥95.0% (by GC)
N-(2-Pyridylmethyl)glycine ethyl ester
分子式: C₁₀H₁₄N₂O₂
分子量: 194.23 g/mol
MDL编号: MFCD00191573
CAS号: 62402-24-8

现在订购

产品技术规格

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR