Tri-p-tolyl phosphite
供应商: TCI
|
Warning
|
Synonyms:
Tris(4-methylphenyl) Phosphite, Phosphorous Acid Tri-p-cresyl Ester, Tri-p-cresyl Phosphite, Phosphorous Acid Tris(4-methylphenyl) Ester, Phosphorous Acid Tri-p-tolyl Ester
P1175-25G
P1175-500G
TCIAP1175-25GEA
0
CNY
TCIAP1175-25G
TCIAP1175-500G
Tri-p-tolyl phosphite
Tri-p-tolyl phosphite
分子式:
C₂₁H₂₁O₃P 分子量: 352.37 g/mol 闪点: 217°C |
MDL编号:
MFCD00041909 CAS号: 620-42-8 EINECS: 210-639-9 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...