Tris(2-ethylhexyl)phosphite ≥90.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Phosphorous Acid Tris(2-ethylhexyl) Ester, Phosphorous Acid Trioctyl Ester, Trioctyl Phosphite
P1169-25ML
P1169-500ML
TCIAP1169-25MLEA
0
CNY
TCIAP1169-25ML
TCIAP1169-500ML
Tris(2-ethylhexyl)phosphite ≥90.0% (by GC)
Tris(2-ethylhexyl)phosphite
分子式:
C₂₄H₅₁O₃P 分子量: 418.64 g/mol 闪点: 194°C |
MDL编号:
MFCD00015266 CAS号: 301-13-3 EINECS: 206-111-2 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...