4-苯基脲唑 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 4-苯基-1,2,4-三唑烷-3,5-二酮

P1160-25G P1160-5G
TCIAP1160-25GEA 0 CNY
TCIAP1160-25G TCIAP1160-5G
4-苯基脲唑 ≥98.0% (by titrimetric analysis)
4-苯基脲唑
分子式: C₈H₇N₃O₂
MDL编号: MFCD00005226
CAS号: 15988-11-1
EINECS: 240-127-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 207.0 to 211.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR