2-Pentynyl chloride ≥96.0% (通过GC)
供应商: TCI
|
Warning
|
P1141-25ML
P1141-5ML
TCIAP1141-25MLEA
0
CNY
TCIAP1141-25ML
TCIAP1141-5ML
2-Pentynyl chloride ≥96.0% (通过GC)
2-Pentynyl chloride
分子式:
C₅H₇Cl 沸点: 122°C 闪点: 25°C |
MDL编号:
MFCD00191678 CAS号: 22592-15-0 EINECS: 245-112-2 UN号: 1993 ADR: 3,III |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...