N,N′-(1,3-亚苯基)二马来酰亚胺 ≥97.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
N,N'-1,3-Bismaleimidobenzene, N,N'-1,3-Phenylenebismaleimide
P0976-25G
P0976-500G
TCIAP0976-25GEA
0
CNY
TCIAP0976-25G
TCIAP0976-500G
N,N′-(1,3-亚苯基)二马来酰亚胺 ≥97.0% (by GC)
N,N′-(1,3-亚苯基)二马来酰亚胺
分子式:
C₁₄H₈N₂O₄ 分子量: 268.23 g/mol |
MDL编号:
MFCD00022569 CAS号: 3006-93-7 EINECS: 221-112-8 UN号: 2811 ADR: 6.1 II |
产品技术规格
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 200.0 to 204.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...