吡啶-2,6-二甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

P0949-1G P0949-5G
TCIAP0949-1GEA 0 CNY
TCIAP0949-1G TCIAP0949-5G
吡啶-2,6-二甲醛 ≥98.0% (by GC)
吡啶-2,6-二甲醛
分子式: C₇H₅NO₂
MDL编号: MFCD00010103
CAS号: 5431-44-7
EINECS: 226-589-6

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR