Sodium monododecyl phosphate (mixture of mono and disodium salt) ≥75.0% (by GC)

供应商: TCI

Synonyms: Phosphoric Acid Monododecyl Ester Sodium Salt, Sodium Monolauryl Phosphate

P0865-25G P0865-500G
TCIAP0865-25GEA 0 CNY
TCIAP0865-25G TCIAP0865-500G
Sodium monododecyl phosphate (mixture of mono and disodium salt) ≥75.0% (by GC)
Sodium monododecyl phosphate (mixture of mono and disodium salt)
MDL编号: MFCD00058971
CAS号: 17026-83-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 75.0 %
Neutralization Value max. 190.0
Sodium Didodecyl Phosphate (GC) max. 10.0 %
Water max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR