Isopropyl propyl sulphide ≥95.0% (by GC)
供应商: TCI
|
Warning
|
P0826-25ML
P0826-5ML
TCIAP0826-25MLEA
0
CNY
TCIAP0826-25ML
TCIAP0826-5ML
Isopropyl propyl sulphide ≥95.0% (by GC)
Isopropyl propyl sulphide
分子式:
C₆H₁₄S 分子量: 118.24 g/mol 沸点: 132°C |
MDL编号:
MFCD00027125 CAS号: 5008-73-1 EINECS: 225-686-0 UN号: 1993 ADR: 3 III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...